Please login to your online account to display your discounted pricing

2-Chloro-5-methylpyrimidine, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-methylpyrimidine
Assay Percent Range 95%
Molecular Formula C5H5ClN2
Formula Weight 128.56
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC454080010

  SDS
thermo fisher scientific
AC454080010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC454082500

 
thermo fisher scientific
AC454082500
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.56
Quantity 1g
Assay Percent Range 95%
Chemical Name or Material 2-Chloro-5-methylpyrimidine
Molecular Formula C5H5ClN2
CAS 22536-61-4