missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-methylpyrimidine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC454080010

 View more versions of this product

Catalog No. AC454080010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Chloro-5-methylpyrimidine
94°C to 95°C
22536-61-4
C5H5ClN2
APRMCBSTMFKLEI-UHFFFAOYSA-N
2-chloro-5-methylpyrimidine
581719
95%
Glass Bottle
1g
94% min. (GC)
2-chloro-5-methyl pyrimidine, 2-chloro-5-methyl-pyrimidine, ksc497i7j, pubchem11095, pyrimidine, 2-chloro-5-methyl
CC1=CN=C(N=C1)Cl
128.56
128.56

Chemical Identifiers

22536-61-4
128.56
2-chloro-5-methyl pyrimidine, 2-chloro-5-methyl-pyrimidine, ksc497i7j, pubchem11095, pyrimidine, 2-chloro-5-methyl
2-chloro-5-methylpyrimidine
C5H5ClN2
APRMCBSTMFKLEI-UHFFFAOYSA-N
581719
CC1=CN=C(N=C1)Cl
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents