missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-5-methylphenol, 99%, ACROS Organics™

$65.74 - $223.97

Chemical Identifiers

CAS 615-74-7
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C7H7ClO
Linear Formula ClC6H3(CH3)OH
Formula Weight 142.58
MDL Number MFCD00002175
Chemical Name or Material 2-chloro-5-methylphenol
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC170415000 SDS Thermo Fisher Scientific
AC170415000
Glass bottle 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC170410250 SDS Thermo Fisher Scientific
170410250
Glass bottle 25g Each for $65.74
AC170411000 SDS Thermo Fisher Scientific
170411000
Glass bottle 100g Each for $223.97
Specifications Description & Specifications

Chemical Identifiers

615-74-7
100.0
C7H7ClO
142.58
2-chloro-5-methylphenol
98.5
99%
ClC6H3(CH3)OH
MFCD00002175
≥98.5%

Specifications

615-74-7
100.0
Glass bottle
45° to 48°C
Beige
500g
ClC6H3(CH3)OH
MFCD00002175
4-chloro-3-hydroxytoluene
≥98.5%
98.5
Authentic
1.215g/mL
Crystalline Powder
99%
C7H7ClO
142.58
06, I, 187
2-chloro-5-methylphenol

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning