missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-5-methylphenol, 99%, ACROS Organics™

Manufacturer:  Acros Organics 170410250

 View more versions of this product

Catalog No. AC170410250



Specifications

Specifications

2-chloro-5-methylphenol
Authentic
196.0°C
45.0°C to 48.0°C
615-74-7
100.0
C7H7ClO
MFCD00002175
6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, 6-chloro-m-cresol, dsstox_cid_26558, dsstox_rid_81718
CC1=CC(=C(C=C1)Cl)O
142.582
142.58
99%
81°C
Glass bottle
Beige
25g
98.5
99%
ClC6H3(CH3)OH
06, I, 187
SMFHPCZZAAMJJO-UHFFFAOYSA-N
2-chloro-5-methylphenol
12008
Crystalline Powder

Chemical Identifiers

615-74-7
142.582
SMFHPCZZAAMJJO-UHFFFAOYSA-N
12008
CC1=CC(=C(C=C1)Cl)O
C7H7ClO
MFCD00002175
6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, 6-chloro-m-cresol, dsstox_cid_26558, dsstox_rid_81718
2-chloro-5-methylphenol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents