Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-methylbenzoic acid, 98%

$120.23 - $1,780.99

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-methylbenzoic acid
CAS 6342-60-5
Molecular Formula C8H7ClO2
Formula Weight 170.6
MDL Number MFCD00045798
Synonym 6-Chloro-m-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2534703

  SDS
alfa aesar™
B2534703
1g Each for $120.23

AAB2534706

  SDS
alfa aesar™
B2534706
5g Each for $449.01

AAB2534714

  SDS
alfa aesar™
B2534714
25g Each for $1,780.99
Description & Specifications

Specifications

Formula Weight 170.6
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C8H7ClO2
CAS 6342-60-5
Chemical Name or Material 2-Chloro-5-methylbenzoic acid
Quantity 1g
MDL Number MFCD00045798
Synonym 6-Chloro-m-toluic acid
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.