missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-methylbenzeneboronic acid, 96%

Manufacturer:  Alfa Aesar™ H5316606

 View more versions of this product

Catalog No. AAH5316606



Specifications

Specifications

2-Chloro-5-methylbenzeneboronic acid
5g
C7H8BClO2
2-chloro-5-methylphenyl boronic acid, 2-chloro-5-methylbenzeneboronic acid, 2-chloro-5-methyl-phenyl boronic acid, boronic acid, 2-chloro-5-methylphenyl, 2-chloranyl-5-methyl-phenyl boronic acid, pubchem23777, acmc-209dni, 6-chloro-m-tolylboronic acid, ksc174k0p, 2-chloro-5-methylphenylboronic
B(C1=C(C=CC(=C1)C)Cl)(O)O
170.399
170.4
203°C to 205°C
193353-35-4
MFCD06659822
JMUXNVYJDBCLPF-UHFFFAOYSA-N
(2-chloro-5-methylphenyl)boronic acid
17750235
96%

Chemical Identifiers

193353-35-4
170.399
JMUXNVYJDBCLPF-UHFFFAOYSA-N
17750235
B(C1=C(C=CC(=C1)C)Cl)(O)O
C7H8BClO2
MFCD06659822
2-chloro-5-methylphenyl boronic acid, 2-chloro-5-methylbenzeneboronic acid, 2-chloro-5-methyl-phenyl boronic acid, boronic acid, 2-chloro-5-methylphenyl, 2-chloranyl-5-methyl-phenyl boronic acid, pubchem23777, acmc-209dni, 6-chloro-m-tolylboronic acid, ksc174k0p, 2-chloro-5-methylphenylboronic
(2-chloro-5-methylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents