Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-methoxybenzimidazole, 94%

$95.28 - $95.28

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-methoxybenzimidazole
CAS 15965-54-5
Molecular Formula C8H7ClN2O
Formula Weight 182.61
MDL Number MFCD00051945
Beilstein 777455
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL08440MD

 
alfa aesar™
L08440MD
250mg Each for $95.28
Description & Specifications

Specifications

Formula Weight 182.61
Chemical Name or Material 2-Chloro-5-methoxybenzimidazole
Assay 94%
Molecular Formula C8H7ClN2O
CAS 15965-54-5
Health Hazard 1 Warning
MDL Number MFCD00051945
Beilstein 777455
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 165° to 168°C
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.