missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-methoxybenzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5335906

 View more versions of this product

Catalog No. AAH5335906



Specifications

Specifications

2-Chloro-5-methoxybenzeneboronic acid
5g
C7H8BClO3
2-chloro-5-methoxyphenyl boronic acid, 2-chloro-5-methoxyphenyl boronic acid, 2-chloro-5-methoxyphenylboronicacid, 3-borono-4-chloroanisole, 2-chloro-5-methoxybenzeneboronic acid, 2-chloro-5-methoxy-phenyl boronic acid, boronic acid, 2-chloro-5-methoxyphenyl, pubchem6932, acmc-209r2h, 2-chloro-5-methoxylphenylboronic acid
B(C1=C(C=CC(=C1)OC)Cl)(O)O
186.398
186.4
106°C to 110°C
89694-46-2
MFCD06659858
REFXAANPQCJZRY-UHFFFAOYSA-N
(2-chloro-5-methoxyphenyl)boronic acid
17750233
98%

Chemical Identifiers

89694-46-2
186.398
REFXAANPQCJZRY-UHFFFAOYSA-N
17750233
B(C1=C(C=CC(=C1)OC)Cl)(O)O
C7H8BClO3
MFCD06659858
2-chloro-5-methoxyphenyl boronic acid, 2-chloro-5-methoxyphenyl boronic acid, 2-chloro-5-methoxyphenylboronicacid, 3-borono-4-chloroanisole, 2-chloro-5-methoxybenzeneboronic acid, 2-chloro-5-methoxy-phenyl boronic acid, boronic acid, 2-chloro-5-methoxyphenyl, pubchem6932, acmc-209r2h, 2-chloro-5-methoxylphenylboronic acid
(2-chloro-5-methoxyphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents