Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-methoxyaniline hydrochloride, 99%

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-methoxyaniline hydrochloride
CAS 85006-21-9
Molecular Formula C7H8ClNO·HCl
Formula Weight 194.06
MDL Number MFCD00012962
UN Number UN2811
Beilstein 3699386
Synonym 6-Chloro-m-anisidine hydrochloride; 3-Amino-4-chloroanisole hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1349003

  SDS
alfa aesar™
L1349003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1349006

  SDS
alfa aesar™
L1349006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1349014

  SDS
alfa aesar™
L1349014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.06
Health Hazard 3 GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Chemical Name or Material 2-Chloro-5-methoxyaniline hydrochloride
Assay 99%
Health Hazard 2 GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
UN Number UN2811
MDL Number MFCD00012962
Molecular Formula C7H8ClNO·HCl
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 1g
Synonym 6-Chloro-m-anisidine hydrochloride; 3-Amino-4-chloroanisole hydrochloride
Beilstein 3699386
Melting Point ≈207°C
CAS 85006-21-9
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.