missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-methoxyaniline hydrochloride, 99%

Manufacturer:  Alfa Aesar™ L1349003

 View more versions of this product

Catalog No. AAL1349003



Specifications

Specifications

2-Chloro-5-methoxyaniline hydrochloride
1g
C7H9Cl2NO
UN2811
2-chloro-5-methoxyaniline hydrochloride, 3-amino-4-chloroanisole hydrochloride, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxyaniline hcl, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, benzenamine, 2-chloro-5-methoxy-, hydrochloride, timtec-bb sbb008038, pubchem3658, pubchem3660
COC1=CC(=C(C=C1)Cl)N.Cl
194.055
194.06
~207°C
85006-21-9
MFCD00012962
3699386
NQWBPXKJBZYGHZ-UHFFFAOYSA-N
2-chloro-5-methoxyaniline;hydrochloride
2723900
99%

Chemical Identifiers

85006-21-9
194.055
NQWBPXKJBZYGHZ-UHFFFAOYSA-N
2723900
COC1=CC(=C(C=C1)Cl)N.Cl
C7H9Cl2NO
MFCD00012962
2-chloro-5-methoxyaniline hydrochloride, 3-amino-4-chloroanisole hydrochloride, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxyaniline hcl, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, benzenamine, 2-chloro-5-methoxy-, hydrochloride, timtec-bb sbb008038, pubchem3658, pubchem3660
2-chloro-5-methoxyaniline;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents