Please login to your online account to display your discounted pricing

2-chloro-5-isocyanatopyridine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-isocyanatopyridine
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H3ClN2O
Formula Weight 154.56
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07678CB

  SDS
fisher scientific company
MO07678CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.56
Quantity 250mg
Assay 97%
CAS 125117-96-6
Assay Percent Range 97%
Chemical Name or Material 2-Chloro-5-isocyanatopyridine
Molecular Formula C6H3ClN2O