Please login to your online account to display your discounted pricing

2-chloro-5-isocyanatopyridine, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  MO07678CB

 View more versions of this product

Catalog No. MO07678CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 154.56
Quantity 250mg
Assay 97%
CAS 125117-96-6
Assay Percent Range 97%
Chemical Name or Material 2-Chloro-5-isocyanatopyridine
Molecular Formula C6H3ClN2O

Specifications

Chemical Name or Material 2-Chloro-5-isocyanatopyridine
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H3ClN2O
Formula Weight 154.56