missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-iodopyrimidine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC454070010

 View more versions of this product

Catalog No. AC454070010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Chloro-5-iodopyrimidine
135°C to 136°C
32779-38-7
C4H2ClIN2
WSZRCNZXKKTLQE-UHFFFAOYSA-N
2-chloro-5-iodopyrimidine
1714223
95%
Glass Bottle
1g
94% min. (GC)
2-chloro-5-iodo-pyrimidine, 2-cloro-5-iodopyrimidine, 5-iodo-2-chloropyrimidine, acmc-209hvo, ksc569e7p, pubchem21488, pyrimidine, 2-chloro-5-iodo, pyrimidine,2-chloro-5-iodo
C1=C(C=NC(=N1)Cl)I
240.43
240.43

Chemical Identifiers

32779-38-7
240.43
2-chloro-5-iodo-pyrimidine, 2-cloro-5-iodopyrimidine, 5-iodo-2-chloropyrimidine, acmc-209hvo, ksc569e7p, pubchem21488, pyrimidine, 2-chloro-5-iodo, pyrimidine,2-chloro-5-iodo
2-chloro-5-iodopyrimidine
C4H2ClIN2
WSZRCNZXKKTLQE-UHFFFAOYSA-N
1714223
C1=C(C=NC(=N1)Cl)I
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents