missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-iodopyrimidine, 95%, ACROS Organics™

Chemical Identifiers

CAS 32779-38-7
Molecular Formula C4H2ClIN2
Molecular Weight (g/mol) 240.43
InChI Key WSZRCNZXKKTLQE-UHFFFAOYSA-N
Synonym 2-chloro-5-iodo-pyrimidine, 2-cloro-5-iodopyrimidine, 5-iodo-2-chloropyrimidine, acmc-209hvo, ksc569e7p, pubchem21488, pyrimidine, 2-chloro-5-iodo, pyrimidine,2-chloro-5-iodo
PubChem CID 1714223
IUPAC Name 2-chloro-5-iodopyrimidine
SMILES C1=C(C=NC(=N1)Cl)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC454072500
SDS View Documents
Thermo Fisher Scientific
AC454072500
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC454070010
SDS View Documents
Thermo Fisher Scientific
AC454070010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

32779-38-7
240.43
2-chloro-5-iodo-pyrimidine, 2-cloro-5-iodopyrimidine, 5-iodo-2-chloropyrimidine, acmc-209hvo, ksc569e7p, pubchem21488, pyrimidine, 2-chloro-5-iodo, pyrimidine,2-chloro-5-iodo
2-chloro-5-iodopyrimidine
C4H2ClIN2
WSZRCNZXKKTLQE-UHFFFAOYSA-N
1714223
C1=C(C=NC(=N1)Cl)I

Specifications

135°C to 136°C
32779-38-7
C4H2ClIN2
WSZRCNZXKKTLQE-UHFFFAOYSA-N
2-chloro-5-iodopyrimidine
1714223
95%
250mg
94% min. (GC)
2-chloro-5-iodo-pyrimidine, 2-cloro-5-iodopyrimidine, 5-iodo-2-chloropyrimidine, acmc-209hvo, ksc569e7p, pubchem21488, pyrimidine, 2-chloro-5-iodo, pyrimidine,2-chloro-5-iodo
C1=C(C=NC(=N1)Cl)I
240.43
240.43
2-Chloro-5-iodopyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS