missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-5-iodopyridine 98.0+%, TCI America™
$121.11 - $361.67
Chemical Identifiers
CAS | 69045-79-0 |
---|---|
Molecular Formula | C5H3ClIN |
Molecular Weight (g/mol) | 239.44 |
MDL Number | MFCD01863635 |
InChI Key | QWLGCWXSNYKKDO-UHFFFAOYSA-N |
Synonym | 2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine |
PubChem CID | 4397176 |
IUPAC Name | 2-chloro-5-iodopyridine |
SMILES | C1=CC(=NC=C1I)Cl |
Chemical Identifiers
69045-79-0 | |
239.44 | |
QWLGCWXSNYKKDO-UHFFFAOYSA-N | |
4397176 | |
C1=CC(=NC=C1I)Cl |
C5H3ClIN | |
MFCD01863635 | |
2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine | |
2-chloro-5-iodopyridine |
Specifications
69045-79-0 | |
Yellow | |
MFCD01863635 | |
2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine | |
C1=CC(=NC=C1I)Cl | |
239.44 | |
239.44 | |
Crystalline Powder |
99°C | |
C5H3ClIN | |
5 g | |
QWLGCWXSNYKKDO-UHFFFAOYSA-N | |
2-chloro-5-iodopyridine | |
4397176 | |
≥98.0% (GC) | |
2-Chloro-5-iodopyridine |
Safety and Handling
TSCA : No