Learn More
2-Chloro-5-iodopyridine, 98%, Thermo Scientific Chemicals
$91.67 - $1,622.67
Chemical Identifiers
CAS | 69045-79-0 |
---|---|
Molecular Formula | C5H3ClIN |
Molecular Weight (g/mol) | 239.44 |
MDL Number | MFCD01863635 |
InChI Key | QWLGCWXSNYKKDO-UHFFFAOYSA-N |
Synonym | 2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine |
PubChem CID | 4397176 |
IUPAC Name | 2-chloro-5-iodopyridine |
SMILES | C1=CC(=NC=C1I)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2527103
|
Thermo Scientific Chemicals
B2527103 |
1 g |
N/A
|
|
|||||
AAB2527106
|
Thermo Scientific Chemicals
B2527106 |
5 g |
N/A
|
|
|||||
AAB2527114
|
Thermo Scientific Chemicals
B2527114 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69045-79-0 | |
239.44 | |
QWLGCWXSNYKKDO-UHFFFAOYSA-N | |
4397176 | |
C1=CC(=NC=C1I)Cl |
C5H3ClIN | |
MFCD01863635 | |
2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine | |
2-chloro-5-iodopyridine |
Specifications
69045-79-0 | |
C5H3ClIN | |
1 g | |
Light sensitive | |
QWLGCWXSNYKKDO-UHFFFAOYSA-N | |
2-chloro-5-iodopyridine | |
4397176 | |
98% |
95°C to 98°C | |
MFCD01863635 | |
108889 | |
2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine | |
C1=CC(=NC=C1I)Cl | |
239.44 | |
239.44 | |
2-Chloro-5-iodopyridine |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only