Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-iodo-3-(trifluoromethyl)pyridine, 95%

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-iodo-3-(trifluoromethyl)pyridine
CAS 887707-25-7
Formula Weight 307.44
MDL Number MFCD12024302
Beilstein 10249610
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6432903

 
alfa aesar™
H6432903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64329MD

 
alfa aesar™
H64329MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6432906

 
alfa aesar™
H6432906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 307.44
Chemical Name or Material 2-Chloro-5-iodo-3-(trifluoromethyl)pyridine
Physical Form Solid
Assay 95%
MDL Number MFCD12024302
CAS 887707-25-7
Beilstein 10249610
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Light Sensitive
Melting Point 43°C
Quantity 1g

Applications
It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.