Learn More
2-Chloro-5-iodo-3-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6432903
Description
It is used as pharmaceutical intermediate and as an advanced chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate and as an advanced chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light.
Specifications
2-Chloro-5-iodo-3-(trifluoromethyl)pyridine | |
43°C | |
MFCD12024302 | |
10249610 | |
2-chloro-5-iodo-3-trifluoromethyl pyridine, 2-chloro-3-trifluoromethyl-5-iodo-pyridine, 2-chloro-3-trifluoromethyl-5-iodopyridine, 2-choro-5-iodo-3-trifluoromethylpyridine, 2-chloro-3-trifluoromethyl-5-iodo pyridine | |
KUASPTUGCBGXAE-UHFFFAOYSA-N | |
2-chloro-5-iodo-3-(trifluoromethyl)pyridine | |
11500513 | |
95% |
887707-25-7 | |
C6H2ClF3IN | |
1 g | |
Light sensitive | |
Slightly soluble in water. | |
FC(F)(F)C1=CC(I)=CN=C1Cl | |
307.44 | |
307.44 | |
Solid |
Chemical Identifiers
887707-25-7 | |
307.44 | |
KUASPTUGCBGXAE-UHFFFAOYSA-N | |
11500513 | |
FC(F)(F)C1=CC(I)=CN=C1Cl |
C6H2ClF3IN | |
MFCD12024302 | |
2-chloro-5-iodo-3-trifluoromethyl pyridine, 2-chloro-3-trifluoromethyl-5-iodo-pyridine, 2-chloro-3-trifluoromethyl-5-iodopyridine, 2-choro-5-iodo-3-trifluoromethylpyridine, 2-chloro-3-trifluoromethyl-5-iodo pyridine | |
2-chloro-5-iodo-3-(trifluoromethyl)pyridine |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only