Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-hydroxybenzoic acid, 99%

$106.66 - $264.76

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-hydroxybenzoic acid
CAS 56961-30-9
Molecular Formula C7H5ClO3
Formula Weight 172.57
MDL Number MFCD04038818
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2611203

  SDS
alfa aesar™
H2611203
1g Each for $264.76

AAH26112MD

  SDS
alfa aesar™
H26112MD
250mg Each for $106.66
Description & Specifications

Specifications

Formula Weight 172.57
Chemical Name or Material 2-Chloro-5-hydroxybenzoic acid
Assay 99%
Molecular Formula C7H5ClO3
MDL Number MFCD04038818
CAS 56961-30-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 178° to 180°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.