missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-hydroxy-3-methylpyridine, 97%

$141.11 - $516.61

Chemical Identifiers

CAS 54232-03-0
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD11519295
InChI Key UGDSWVXJJFIVAH-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxy-3-methylpyridine, 2-chloro-5-hydroxy-3-methylpyrdine, 6-chloro-5-methyl-pyridin-3-ol, 3-pyridinol, 6-chloro-5-methyl, 2-chloro-5-hydroxy-3-picoline, 6-chloro-5-methyl-3-pyridinol, 2-chloro-3-methyl-5-hydroxypyridine, pubchem12478, acmc-209len, 6-chloro-5-methylpyridine-3-ol
PubChem CID 12852015
IUPAC Name 6-chloro-5-methylpyridin-3-ol
SMILES CC1=CC(=CN=C1Cl)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64986MD View Documents Alfa Aesar™
H64986MD
250mg Each for $141.11
 
AAH6498603 View Documents Alfa Aesar™
H6498603
1g Each for $516.61
 
Specifications

Chemical Identifiers

54232-03-0
143.57
UGDSWVXJJFIVAH-UHFFFAOYSA-N
12852015
CC1=CC(=CN=C1Cl)O
C6H6ClNO
MFCD11519295
2-chloro-5-hydroxy-3-methylpyridine, 2-chloro-5-hydroxy-3-methylpyrdine, 6-chloro-5-methyl-pyridin-3-ol, 3-pyridinol, 6-chloro-5-methyl, 2-chloro-5-hydroxy-3-picoline, 6-chloro-5-methyl-3-pyridinol, 2-chloro-3-methyl-5-hydroxypyridine, pubchem12478, acmc-209len, 6-chloro-5-methylpyridine-3-ol
6-chloro-5-methylpyridin-3-ol

Specifications

250mg
C6H6ClNO
2-chloro-5-hydroxy-3-methylpyridine, 2-chloro-5-hydroxy-3-methylpyrdine, 6-chloro-5-methyl-pyridin-3-ol, 3-pyridinol, 6-chloro-5-methyl, 2-chloro-5-hydroxy-3-picoline, 6-chloro-5-methyl-3-pyridinol, 2-chloro-3-methyl-5-hydroxypyridine, pubchem12478, acmc-209len, 6-chloro-5-methylpyridine-3-ol
CC1=CC(=CN=C1Cl)O
143.57
143.57
2-Chloro-5-hydroxy-3-methylpyridine
54232-03-0
MFCD11519295
UGDSWVXJJFIVAH-UHFFFAOYSA-N
6-chloro-5-methylpyridin-3-ol
12852015
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning