missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-(fluorosulfonyl)benzoic acid, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 162190250

 View more versions of this product

Catalog No. AC162190250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Chloro-5-(fluorosulfonyl)benzoic acid
White to slightly brown
25g
21346-66-7
Glass bottle
Crystalline Powder
99+%
ClC6H3(SO2F)CO2H
MFCD00007418
Authentic
148° to 151°C
99+%
99.0
100.0
≥99.0%
C7H4ClFO4S
238.62

Chemical Identifiers

21346-66-7
100.0
C7H4ClFO4S
238.62
2-Chloro-5-(fluorosulfonyl)benzoic acid, 99+%
99.0
99+%
ClC6H3(SO2F)CO2H
MFCD00007418
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 244-338-9

TSCA : TSCA