Please login to your online account to display your discounted pricing

2-Chloro-5-(fluorosulfonyl)benzoic acid, 99+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  162190050

 View more versions of this product

Catalog No. AC162190050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 238.62
Assay ≥99.0%
Physical Form Crystalline Powder
Quantity 5g
Color White to slightly brown
TSCA TSCA
EINECS Number 244-338-9
Melting Point 148° to 151°C
CAS Max % 100.0
CAS Min % 99.0
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Linear Formula ClC6H3(SO2F)CO2H
CAS 21346-66-7
Name Note 99+%
Molecular Formula C7H4ClFO4S
Chemical Name or Material 2-Chloro-5-(fluorosulfonyl)benzoic acid
MDL Number MFCD00007418
Assay Percent Range 99+%
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Chloro-5-(fluorosulfonyl)benzoic acid
Name Note 99+%
CAS 21346-66-7
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H4ClFO4S
Linear Formula ClC6H3(SO2F)CO2H
Formula Weight 238.62
MDL Number MFCD00007418
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents