Please login to your online account to display your discounted pricing

2-Chloro-5-(fluorosulfonyl)benzoic acid, 99+%, ACROS Organics™

$45.22 - $45.22

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-(fluorosulfonyl)benzoic acid
Name Note 99+%
CAS 21346-66-7
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H4ClFO4S
Linear Formula ClC6H3(SO2F)CO2H
Formula Weight 238.62
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC162190250

  SDS
thermo fisher scientific
162190250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC162190050

  SDS
thermo fisher scientific
162190050
Glass bottle 5g Each for $45.22
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 238.62
Quantity 25g
Color White to slightly brown
EINECS Number 244-338-9
TSCA TSCA
Melting Point 148° to 151°C
CAS Min % 99.0
CAS Max % 100.0
Name Note 99+%
Assay Percent Range 99+%
Linear Formula ClC6H3(SO2F)CO2H
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Chemical Name or Material 2-Chloro-5-(fluorosulfonyl)benzoic acid
Molecular Formula C7H4ClFO4S
MDL Number MFCD00007418
CAS 21346-66-7
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.