missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-fluoropyrimidine, 97%

Manufacturer:  Alfa Aesar™ H55055MD

 View more versions of this product

Catalog No. AAH55055MD



Description

Description

Applications
2-Chloro-5-fluoropyrimidine is used to prepare 2,4-disubstituted-5-fluoropyrimidine , which is a biologically active molecule seen in anticancer agents like 5-fluorouracil.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

2-Chloro-5-fluoropyrimidine
65°C (149°F)
250mg
C4H2ClFN2
UN3267
AGYUQBNABXVWMS-UHFFFAOYSA-N
2-chloro-5-fluoropyrimidine
583466
97%
1.073
1.503
62802-42-0
MFCD03788197
#
C1=C(C=NC(=N1)Cl)F
132.522
132.52

Chemical Identifiers

62802-42-0
132.522
AGYUQBNABXVWMS-UHFFFAOYSA-N
583466
C1=C(C=NC(=N1)Cl)F
C4H2ClFN2
MFCD03788197
#
2-chloro-5-fluoropyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents