missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-fluoropyridine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442860010

 View more versions of this product

Catalog No. AC442860010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Chloro-5-fluoropyridine
42°C
1.297
23°C to 24°C
31301-51-6
C5H3ClFN
QOGXQLSFJCIDNY-UHFFFAOYSA-N
2-chloro-5-fluoropyridine
2782801
95%
1.2970g/mL
Glass Bottle
74°C (37.0 Torr)
1g
94% min. (GC)
2-chlor-5-fluorpyridin, 2-chloro-5-fluoro pyridine, 2-chloro-5-fluoro-pyridine, 2-chloro-5-fluropyridine, acmc-1aevz, ksc223e3t, pubchem1097, pyridine, 2-chloro-5-fluoro, tpc-py082, zlchem 338
C1=CC(=NC=C1F)Cl
131.54
131.54

Chemical Identifiers

31301-51-6
131.54
2-chlor-5-fluorpyridin, 2-chloro-5-fluoro pyridine, 2-chloro-5-fluoro-pyridine, 2-chloro-5-fluropyridine, acmc-1aevz, ksc223e3t, pubchem1097, pyridine, 2-chloro-5-fluoro, tpc-py082, zlchem 338
2-chloro-5-fluoropyridine
C5H3ClFN
QOGXQLSFJCIDNY-UHFFFAOYSA-N
2782801
C1=CC(=NC=C1F)Cl
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS Documents