Please login to your online account to display your discounted pricing

2-Chloro-5-fluoropyridine, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-fluoropyridine
Assay Percent Range 95%
Molecular Formula C5H3ClFN
Formula Weight 131.54
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC442860050

 
thermo fisher scientific
AC442860050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442860010

 
thermo fisher scientific
AC442860010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 131.54
Quantity 5g
CAS 31301-51-6
Chemical Name or Material 2-Chloro-5-fluoropyridine
Assay Percent Range 95%
Molecular Formula C5H3ClFN