missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-fluorophenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC249960250

 View more versions of this product

Catalog No. AC249960250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Chloro-5-fluorophenol
Glass bottle
183°C to 185°C
25g
97.5
97.5% min. (GC)
FC6H3(OH)Cl
2-chloro-5-fluoro-phenol, 2-chloro-5-fluorphenol, acmc-209iz2, attercop-chm at137231, ksc226k0t, pubchem2146
CMQOZIKIOASEIN-UHFFFAOYSA-N
2-chloro-5-fluorophenol
2724522
Liquid
Authentic
1.5240 to 1.526
Colorless to Yellow
3827-49-4
100.0
C6H4ClFO
MFCD00042524
Solubility in water: slightly soluble. Other solubilities: soluble in aqueous alkali,soluble in most common organic solvents
C1=CC(=C(C=C1F)O)Cl
146.55
146.55
98%

Chemical Identifiers

3827-49-4
146.55
CMQOZIKIOASEIN-UHFFFAOYSA-N
2724522
C1=CC(=C(C=C1F)O)Cl
C6H4ClFO
MFCD00042524
2-chloro-5-fluoro-phenol, 2-chloro-5-fluorphenol, acmc-209iz2, attercop-chm at137231, ksc226k0t, pubchem2146
2-chloro-5-fluorophenol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents