Please login to your online account to display your discounted pricing

2-Chloro-5-fluoronitrobenzene, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-fluoronitrobenzene
Name Note 99+%
CAS 345-17-5
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H3ClFNO2
Linear Formula ClC6H3(F)NO2
Formula Weight 175.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC294060050

  SDS
thermo fisher scientific
294060050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Crystalline Solid
Formula Weight 175.55
Quantity 5g
Color Yellow
EINECS Number 206-456-9
TSCA TSCA
Melting Point 37° to 40°C
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 2-Chloro-5-fluoronitrobenzene
Infrared Spectrum Authentic
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C6H3ClFNO2
Linear Formula ClC6H3(F)NO2
MDL Number MFCD00084853
CAS 345-17-5
Synonym 1-Chloro-4-fluoro-2-nitrobenzene
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.