missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-fluorobenzyl bromide, 98%

Manufacturer:  Alfa Aesar™ H6125906

 View more versions of this product

Catalog No. AAH6125906



Specifications

Specifications

2-Chloro-5-fluorobenzyl bromide
1.5660
Colorless
81778-09-8
MFCD01631419
1-bromomethyl-2-chloro-5-fluorobenzene
C1=CC(=C(C=C1F)CBr)Cl
223.469
200.01
98%
1.654g/mL
65°C to 66°C
5g
C7H5BrClF
UN2924
AUVLFQDKJFSFIX-UHFFFAOYSA-N
2-(bromomethyl)-1-chloro-4-fluorobenzene
2773623
Liquid

Chemical Identifiers

81778-09-8
223.469
AUVLFQDKJFSFIX-UHFFFAOYSA-N
2773623
C1=CC(=C(C=C1F)CBr)Cl
C7H5BrClF
MFCD01631419
1-bromomethyl-2-chloro-5-fluorobenzene
2-(bromomethyl)-1-chloro-4-fluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

Documents