missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-fluorobenzyl alcohol, 97+%

$71.58 - $258.59

Chemical Identifiers

CAS 261762-59-8
Molecular Formula C7H6ClFO
Formula Weight 160.58
MDL Number MFCD01631418
Chemical Name or Material 2-Chloro-5-fluorobenzyl alcohol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2443103 SDS Alfa Aesar™
B2443103
1g Each for $71.58
AAB2443106 SDS Alfa Aesar™
B2443106
5g Each for $258.59
Specifications Description & Specifications

Chemical Identifiers

261762-59-8
160.58
2-Chloro-5-fluorobenzyl alcohol
C7H6ClFO
MFCD01631418

Specifications

68° to 72°C
261762-59-8
160.58
2-Chloro-5-fluorobenzyl alcohol
1g
C7H6ClFO
MFCD01631418
≥97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning