Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-fluorobenzoic acid, 97%

$65.42 - $594.16

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-fluorobenzoic acid
CAS 2252-50-8
Molecular Formula C7H4ClFO2
Formula Weight 174.56
MDL Number MFCD00156967
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2646203

  SDS
alfa aesar™
H2646203
1g Each for $65.42

AAH2646206

  SDS
alfa aesar™
H2646206
5g Each for $193.95

AAH2646214

  SDS
alfa aesar™
H2646214
25g Each for $594.16
Description & Specifications

Specifications

Formula Weight 174.56
Chemical Name or Material 2-Chloro-5-fluorobenzoic acid
Assay 97%
Molecular Formula C7H4ClFO2
MDL Number MFCD00156967
CAS 2252-50-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 147° to 150°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.