Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-fluorobenzamide, 97+%

$64.03 - $64.03

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-fluorobenzamide
CAS 88487-25-6
Molecular Formula C7H5ClFNO
Formula Weight 173.58
MDL Number MFCD01631465
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2309303

 
alfa aesar™
B2309303
1g Each for $64.03
Description & Specifications

Specifications

Formula Weight 173.58
Chemical Name or Material 2-Chloro-5-fluorobenzamide
Assay ≥97%
Molecular Formula C7H5ClFNO
MDL Number MFCD01631465
CAS 88487-25-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.