missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-fluoro-6-methylpyridine, 95%

$38.86 - $154.22

Chemical Identifiers

CAS 884494-78-4
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD04972391
InChI Key ICSRNKKGNHKSJH-UHFFFAOYSA-N
Synonym 2-chloro-5-fluoro-6-methylpyridine, 2-chloro-5-fluoro-6-picoline, 6-chloro-3-fluoro-2-methyl-pyridine, 6-chloro-3-fluoro-2-picoline, pubchem6219, ksc659q3d, pyridine,6-chloro-3-fluoro-2-methyl, 6-chloranyl-3-fluoranyl-2-methyl-pyridine
PubChem CID 19867376
IUPAC Name 6-chloro-3-fluoro-2-methylpyridine
SMILES CC1=C(C=CC(=N1)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6434703 View Documents Alfa Aesar™
H6434703
1g Each for $38.86
 
AAH6434706 View Documents Alfa Aesar™
H6434706
5g Each for $154.22
 
Specifications

Chemical Identifiers

884494-78-4
145.561
ICSRNKKGNHKSJH-UHFFFAOYSA-N
19867376
CC1=C(C=CC(=N1)Cl)F
C6H5ClFN
MFCD04972391
2-chloro-5-fluoro-6-methylpyridine, 2-chloro-5-fluoro-6-picoline, 6-chloro-3-fluoro-2-methyl-pyridine, 6-chloro-3-fluoro-2-picoline, pubchem6219, ksc659q3d, pyridine,6-chloro-3-fluoro-2-methyl, 6-chloranyl-3-fluoranyl-2-methyl-pyridine
6-chloro-3-fluoro-2-methylpyridine

Specifications

1g
C6H5ClFN
13638700
ICSRNKKGNHKSJH-UHFFFAOYSA-N
6-chloro-3-fluoro-2-methylpyridine
19867376
Solid
2-Chloro-5-fluoro-6-methylpyridine
884494-78-4
MFCD04972391
2-chloro-5-fluoro-6-methylpyridine, 2-chloro-5-fluoro-6-picoline, 6-chloro-3-fluoro-2-methyl-pyridine, 6-chloro-3-fluoro-2-picoline, pubchem6219, ksc659q3d, pyridine,6-chloro-3-fluoro-2-methyl, 6-chloranyl-3-fluoranyl-2-methyl-pyridine
CC1=C(C=CC(=N1)Cl)F
145.561
145.56
95%