missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-fluoro-3-nitropyridine, TCI America™

Manufacturer:  TCI America C31151G

 View more versions of this product

Catalog No. C31151G



Specifications

Specifications

2-Chloro-5-fluoro-3-nitropyridine
50°C
136888-21-6
MFCD06659490
SVVZGNAZMPSGMU-UHFFFAOYSA-N
2-chloro-5-fluoro-3-nitropyridine
10773626
Crystal-Powder at 20°C
Yellow
1g
C5H2ClFN2O2
2-chloro-3-nitro-5-fluoropyridine, 2-chloro-5-fluoro-3-nitro-pyridine, pyridine, 2-chloro-5-fluoro-3-nitro, pubchem2967, acmc-1cc17, 2-chloro-5-fluoro-3-nitro pyridine, pyridine,2-chloro-5-fluoro-3-nitro
C1=C(C(=NC=C1F)Cl)[N+](=O)[O-]
176.531
176.53
≥98.0% (GC)

Chemical Identifiers

136888-21-6
176.531
SVVZGNAZMPSGMU-UHFFFAOYSA-N
10773626
C1=C(C(=NC=C1F)Cl)[N+](=O)[O-]
C5H2ClFN2O2
MFCD06659490
2-chloro-3-nitro-5-fluoropyridine, 2-chloro-5-fluoro-3-nitro-pyridine, pyridine, 2-chloro-5-fluoro-3-nitro, pubchem2967, acmc-1cc17, 2-chloro-5-fluoro-3-nitro pyridine, pyridine,2-chloro-5-fluoro-3-nitro
2-chloro-5-fluoro-3-nitropyridine
Safety and Handling

Safety and Handling

TSCA : No

Documents