missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-fluoro-3-methylbenzoic acid, 98%

Manufacturer:  Alfa Aesar™ H6316503

 View more versions of this product

Catalog No. AAH6316503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Chloro-5-fluoro-3-methylbenzoic acid
1g
C8H6ClFO2
2-chloro-5-fluoro-3-methylbenzoicacid
CC1=C(C(=CC(=C1)F)C(=O)O)Cl
188.582
188.58
98%
White
1427416-28-1
MFCD23712098
AILHHXAQZADVIH-UHFFFAOYSA-N
2-chloro-5-fluoro-3-methylbenzoic acid
53430534
Solid

Chemical Identifiers

1427416-28-1
188.582
AILHHXAQZADVIH-UHFFFAOYSA-N
53430534
CC1=C(C(=CC(=C1)F)C(=O)O)Cl
C8H6ClFO2
MFCD23712098
2-chloro-5-fluoro-3-methylbenzoicacid
2-chloro-5-fluoro-3-methylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents