missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-5-ethylpyrimidine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441730010

 View more versions of this product

Catalog No. AC441730010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-chloro-5-ethylpyrimidine
>109°C
Glass bottle
223°C
1g
96.0
96% min. (GC)
2-chloro-5-ethyl pyrimidine, 2-chloro-5-ethyl-pyrimidine, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, 5-ethyl-2-chloropyrimidine, acmc-1brw8, ksc493s1b, pubchem7032, pyrimidine, 2-chloro-5-ethyl
CCC1=CN=C(N=C1)Cl
142.59
142.59
97%
1.1740g/mL
Authentic
1.174
Colorless to Yellow
111196-81-7
100.0
C6H7ClN2
BGLLZQRUXJGTAD-UHFFFAOYSA-N
2-chloro-5-ethylpyrimidine
3572763
Liquid or Low Melting Solid

Chemical Identifiers

111196-81-7
142.59
2-chloro-5-ethyl pyrimidine, 2-chloro-5-ethyl-pyrimidine, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, 5-ethyl-2-chloropyrimidine, acmc-1brw8, ksc493s1b, pubchem7032, pyrimidine, 2-chloro-5-ethyl
2-chloro-5-ethylpyrimidine
C6H7ClN2
BGLLZQRUXJGTAD-UHFFFAOYSA-N
3572763
CCC1=CN=C(N=C1)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents