missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(dimethylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5465503

 View more versions of this product

Catalog No. AAH5465503



Specifications

Specifications

2-Chloro-5-(dimethylcarbamoyl)benzeneboronic acid
1449144-75-5
MFCD22125080
LVOHUMGEBQAIBQ-UHFFFAOYSA-N
[2-chloro-5-(dimethylcarbamoyl)phenyl]boronic acid
99738090
97%
1g
C9H11BClNO3
2-Chloro-5-(dimethylcarbamoyl)phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)N(C)C)Cl)(O)O
227.451
227.45

Chemical Identifiers

1449144-75-5
227.451
LVOHUMGEBQAIBQ-UHFFFAOYSA-N
99738090
B(C1=C(C=CC(=C1)C(=O)N(C)C)Cl)(O)O
C9H11BClNO3
MFCD22125080
2-Chloro-5-(dimethylcarbamoyl)phenylboronic acid
[2-chloro-5-(dimethylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents