missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(cyclopropylcarbamoyl)benzeneboronic acid, 97%

$303.62 - $909.12

Chemical Identifiers

CAS 1449132-64-2
Molecular Formula C10H11BClNO3
Molecular Weight (g/mol) 239.462
MDL Number MFCD22125077
InChI Key ZXCSFHNBVHLHBP-UHFFFAOYSA-N
Synonym 2-chloro-5-cyclopropylcarbamoyl benzeneboronic acid
PubChem CID 73995882
IUPAC Name [2-chloro-5-(cyclopropylcarbamoyl)phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(=O)NC2CC2)Cl)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54576MD View Documents Alfa Aesar™
H54576MD
250mg Each for $303.62
 
AAH5457603 View Documents Alfa Aesar™
H5457603
1g Each for $909.12
 
Description

Description

Applications
2-Chloro-5-(cyclopropylcarbamoyl)benzeneboronic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1449132-64-2
239.462
ZXCSFHNBVHLHBP-UHFFFAOYSA-N
73995882
B(C1=C(C=CC(=C1)C(=O)NC2CC2)Cl)(O)O
C10H11BClNO3
MFCD22125077
2-chloro-5-cyclopropylcarbamoyl benzeneboronic acid
[2-chloro-5-(cyclopropylcarbamoyl)phenyl]boronic acid

Specifications

250mg
C10H11BClNO3
2-chloro-5-cyclopropylcarbamoyl benzeneboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2CC2)Cl)(O)O
239.462
239.46
2-Chloro-5-(cyclopropylcarbamoyl)benzeneboronic acid
1449132-64-2
MFCD22125077
ZXCSFHNBVHLHBP-UHFFFAOYSA-N
[2-chloro-5-(cyclopropylcarbamoyl)phenyl]boronic acid
73995882
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures