missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(cyclopropylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5457603

 View more versions of this product

Catalog No. AAH5457603



Description

Description

Applications
2-Chloro-5-(cyclopropylcarbamoyl)benzeneboronic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-Chloro-5-(cyclopropylcarbamoyl)benzeneboronic acid
1449132-64-2
MFCD22125077
ZXCSFHNBVHLHBP-UHFFFAOYSA-N
[2-chloro-5-(cyclopropylcarbamoyl)phenyl]boronic acid
73995882
97%
1g
C10H11BClNO3
2-chloro-5-cyclopropylcarbamoyl benzeneboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2CC2)Cl)(O)O
239.462
239.46

Chemical Identifiers

1449132-64-2
239.462
ZXCSFHNBVHLHBP-UHFFFAOYSA-N
73995882
B(C1=C(C=CC(=C1)C(=O)NC2CC2)Cl)(O)O
C10H11BClNO3
MFCD22125077
2-chloro-5-cyclopropylcarbamoyl benzeneboronic acid
[2-chloro-5-(cyclopropylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents