Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(chloromethyl)thiophene, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-(chloromethyl)thiophene
CAS 23784-96-5
Formula Weight 167.06
MDL Number MFCD00009764
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5436103

 
alfa aesar™
H5436103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5436106

 
alfa aesar™
H5436106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.06
Boiling Point 83° to 85°C
Chemical Name or Material 2-Chloro-5-(chloromethyl)thiophene
Refractive Index 1.575
Physical Form Liquid
Assay 97%
MDL Number MFCD00009764
CAS 23784-96-5
Storage Note 1 -30° to -10°C
Density 1.385g/mL
Color Colorless to brown
Quantity 1g
Safety and Handling

-30° to -10°C