missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5,5-dimethyl-1,3-cyclohexanedione, 98%

$88.70 - $925.97

Chemical Identifiers

CAS 7298-89-7
Molecular Formula C8H11ClO2
Formula Weight 174.62
MDL Number MFCD00010502
Beilstein 2326749
Chemical Name or Material 2-Chloro-5,5-dimethyl-1,3-cyclohexanedione
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5103503 SDS Alfa Aesar™
H5103503
1g Each for $228.88
AAH5103506 SDS Alfa Aesar™
H5103506
5g Each for $925.97
AAH51035MD SDS Alfa Aesar™
H51035MD
250mg Each for $88.70
Specifications

Description

Applications
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

7298-89-7
174.62
2326749
C8H11ClO2
MFCD00010502
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione

Specifications

161° to 164°C
7298-89-7
174.62
2326749
98%
1g
C8H11ClO2
MFCD00010502
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning