missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(3-fluorobenzylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H55931MD

 View more versions of this product

Catalog No. AAH55931MD



Description

Description

Specifications

Specifications

2-Chloro-5-(3-fluorobenzylcarbamoyl)benzeneboronic acid
1449133-71-4
MFCD20040333
BULLMNJRUBFQST-UHFFFAOYSA-N
[2-chloro-5-[(3-fluorophenyl)methylcarbamoyl]phenyl]boronic acid
73996195
97%
250mg
C14H12BClFNO3
2-chloro-5-3-fluorobenzylcarbamoyl benzeneboronic acid, 2-chloro-5-3-fluorobenzylcarbamoyl phenylboronic acid, 2-chloro-5-3-fluorophenyl methyl carbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NCC2=CC(=CC=C2)F)Cl)(O)O
307.512
307.51

Chemical Identifiers

1449133-71-4
307.512
BULLMNJRUBFQST-UHFFFAOYSA-N
73996195
B(C1=C(C=CC(=C1)C(=O)NCC2=CC(=CC=C2)F)Cl)(O)O
C14H12BClFNO3
MFCD20040333
2-chloro-5-3-fluorobenzylcarbamoyl benzeneboronic acid, 2-chloro-5-3-fluorobenzylcarbamoyl phenylboronic acid, 2-chloro-5-3-fluorophenyl methyl carbamoyl phenylboronic acid
[2-chloro-5-[(3-fluorophenyl)methylcarbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents