missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(2-methylphenylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5562403

 View more versions of this product

Catalog No. AAH5562403



Specifications

Specifications

2-Chloro-5-(2-methylphenylcarbamoyl)benzeneboronic acid
1449132-74-4
MFCD22125160
FNBSKWAKHIKIJI-UHFFFAOYSA-N
[2-chloro-5-[(2-methylphenyl)carbamoyl]phenyl]boronic acid
73996155
97%
1g
C14H13BClNO3
2-chloro-5-2-methylphenyl carbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CC=CC=C2C)Cl)(O)O
289.522
289.53

Chemical Identifiers

1449132-74-4
289.522
FNBSKWAKHIKIJI-UHFFFAOYSA-N
73996155
B(C1=C(C=CC(=C1)C(=O)NC2=CC=CC=C2C)Cl)(O)O
C14H13BClNO3
MFCD22125160
2-chloro-5-2-methylphenyl carbamoyl phenylboronic acid
[2-chloro-5-[(2-methylphenyl)carbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents