missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(2-fluoro-5-methylphenylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5555803

 View more versions of this product

Catalog No. AAH5555803



Specifications

Specifications

2-Chloro-5-(2-fluoro-5-methylphenylcarbamoyl)benzeneboronic acid
2096334-19-7
C14H12BClFNO3
2-Chloro-5-(2-fluoro-5-methylphenylcarbamoyl)phenylboronic acid
307.51
307.51
1g
97%
MFCD20091220
XLACLEABARDMPI-UHFFFAOYSA-N
73996150

Chemical Identifiers

2096334-19-7
307.51
XLACLEABARDMPI-UHFFFAOYSA-N
73996150
C14H12BClFNO3
MFCD20091220
2-Chloro-5-(2-fluoro-5-methylphenylcarbamoyl)phenylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents