missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(2,5-difluorophenylcarbamoyl)benzeneboronic acid, 97%

$303.62 - $909.12

Chemical Identifiers

CAS 1449132-59-5
Molecular Formula C13H9BClF2NO3
Molecular Weight (g/mol) 311.476
MDL Number MFCD20040200
InChI Key OPNXXIVOUWDWMG-UHFFFAOYSA-N
Synonym 2-chloro-5-2,5-difluorophenyl carbamoyl phenylboronic acid
PubChem CID 73995837
IUPAC Name [2-chloro-5-[(2,5-difluorophenyl)carbamoyl]phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)F)F)Cl)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54492MD View Documents Alfa Aesar™
H54492MD
250mg Each for $303.62
 
AAH5449203 View Documents Alfa Aesar™
H5449203
1g Each for $909.12
 
Specifications

Chemical Identifiers

1449132-59-5
311.476
OPNXXIVOUWDWMG-UHFFFAOYSA-N
73995837
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)F)F)Cl)(O)O
C13H9BClF2NO3
MFCD20040200
2-chloro-5-2,5-difluorophenyl carbamoyl phenylboronic acid
[2-chloro-5-[(2,5-difluorophenyl)carbamoyl]phenyl]boronic acid

Specifications

250mg
C13H9BClF2NO3
2-chloro-5-2,5-difluorophenyl carbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)F)F)Cl)(O)O
311.476
311.48
2-Chloro-5-(2,5-difluorophenylcarbamoyl)benzeneboronic acid
1449132-59-5
MFCD20040200
OPNXXIVOUWDWMG-UHFFFAOYSA-N
[2-chloro-5-[(2,5-difluorophenyl)carbamoyl]phenyl]boronic acid
73995837
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures