missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(2,5-dichlorophenylcarbamoyl)benzeneboronic acid, 97%

$279.65 - $837.35

Chemical Identifiers

CAS 1449132-57-3
Molecular Formula C13H9BCl3NO3
Molecular Weight (g/mol) 344.379
MDL Number MFCD22125068
InChI Key LGHQYAUSSYHBTK-UHFFFAOYSA-N
Synonym 2-chloro-5-2,5-dichlorophenyl carbamoyl phenylboronic acid
PubChem CID 73995808
IUPAC Name [2-chloro-5-[(2,5-dichlorophenyl)carbamoyl]phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl)Cl)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54433MD View Documents Alfa Aesar™
H54433MD
250mg Each for $279.65
 
AAH5443303 View Documents Alfa Aesar™
H5443303
1g Each for $837.35
 
Specifications

Chemical Identifiers

1449132-57-3
344.379
LGHQYAUSSYHBTK-UHFFFAOYSA-N
73995808
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl)Cl)(O)O
C13H9BCl3NO3
MFCD22125068
2-chloro-5-2,5-dichlorophenyl carbamoyl phenylboronic acid
[2-chloro-5-[(2,5-dichlorophenyl)carbamoyl]phenyl]boronic acid

Specifications

250mg
C13H9BCl3NO3
2-chloro-5-2,5-dichlorophenyl carbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl)Cl)(O)O
344.379
344.39
2-Chloro-5-(2,5-dichlorophenylcarbamoyl)benzeneboronic acid
1449132-57-3
MFCD22125068
LGHQYAUSSYHBTK-UHFFFAOYSA-N
[2-chloro-5-[(2,5-dichlorophenyl)carbamoyl]phenyl]boronic acid
73995808
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning