missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(2,5-dichlorophenylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5443303

 View more versions of this product

Catalog No. AAH5443303



Specifications

Specifications

2-Chloro-5-(2,5-dichlorophenylcarbamoyl)benzeneboronic acid
1449132-57-3
MFCD22125068
LGHQYAUSSYHBTK-UHFFFAOYSA-N
[2-chloro-5-[(2,5-dichlorophenyl)carbamoyl]phenyl]boronic acid
73995808
97%
1g
C13H9BCl3NO3
2-chloro-5-2,5-dichlorophenyl carbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl)Cl)(O)O
344.379
344.39

Chemical Identifiers

1449132-57-3
344.379
LGHQYAUSSYHBTK-UHFFFAOYSA-N
73995808
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl)Cl)(O)O
C13H9BCl3NO3
MFCD22125068
2-chloro-5-2,5-dichlorophenyl carbamoyl phenylboronic acid
[2-chloro-5-[(2,5-dichlorophenyl)carbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents