missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(2,3-dichlorophenylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5457403

 View more versions of this product

Catalog No. AAH5457403



Specifications

Specifications

2-Chloro-5-(2,3-dichlorophenylcarbamoyl)benzeneboronic acid
1449144-73-3
MFCD27996215
JISRVYPGQJIELD-UHFFFAOYSA-N
[2-chloro-5-[(2,3-dichlorophenyl)carbamoyl]phenyl]boronic acid
73995881
97%
1g
C13H9BCl3NO3
2-chloro-5-2,3-dichlorophenyl carbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C(=CC=C2)Cl)Cl)Cl)(O)O
344.379
344.39

Chemical Identifiers

1449144-73-3
344.379
JISRVYPGQJIELD-UHFFFAOYSA-N
73995881
B(C1=C(C=CC(=C1)C(=O)NC2=C(C(=CC=C2)Cl)Cl)Cl)(O)O
C13H9BCl3NO3
MFCD27996215
2-chloro-5-2,3-dichlorophenyl carbamoyl phenylboronic acid
[2-chloro-5-[(2,3-dichlorophenyl)carbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents