missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-(trifluoromethyl)pyridine, 98+%

Manufacturer:  Alfa Aesar™ L10713MD

 View more versions of this product

Catalog No. AAL10713MD



Specifications

Specifications

2-Chloro-4-(trifluoromethyl)pyridine
145°C
81565-18-6
MFCD00042224
4246276
GBNPVXZNWBWNEN-UHFFFAOYSA-N
2-chloro-4-(trifluoromethyl)pyridine
144958
≥98%
40°C (104°F)
250mg
C6H3ClF3N
UN1992
2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine, 2-chloro-4-trifluoromethyl pyridine
C1=CN=C(C=C1C(F)(F)F)Cl
181.542
181.55

Chemical Identifiers

81565-18-6
181.542
GBNPVXZNWBWNEN-UHFFFAOYSA-N
144958
C1=CN=C(C=C1C(F)(F)F)Cl
C6H3ClF3N
MFCD00042224
2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine, 2-chloro-4-trifluoromethyl pyridine
2-chloro-4-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

Documents