Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-4-(trifluoromethyl)benzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzonitrile
CAS 1813-33-8
Molecular Formula C8H3ClF3N
Formula Weight 205.57
MDL Number MFCD00084939
UN Number UN3276
Synonym 3-Chloro-4-cyanobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2689403

  SDS
alfa aesar™
H2689403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2689406

  SDS
alfa aesar™
H2689406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2689414

  SDS
alfa aesar™
H2689414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.57
Synonym 3-Chloro-4-cyanobenzotrifluoride
Molecular Formula C8H3ClF3N
Boiling Point 192° to 193°C
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzonitrile
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Assay 98%
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Melting Point 82° to 83°C (8mmHg)
MDL Number MFCD00084939
CAS 1813-33-8
Flash Point >104°C (220°F)
Health Hazard 1 Danger
Quantity 1g
Odor Characteristic
UN Number UN3276
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.